missing translation for 'onlineSavingsMsg'
Learn More

Theophylline, 99.51%, MP Biomedicals™

Product Code. 11483411 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483411 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483411 Supplier MP Biomedicals Supplier No. 0210302480
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 58-55-9
Color White
Assay Percent Range 0.9951
Molecular Formula C7H8N4O2
Quantity 100 g
Synonym theophylline, 1,3-dimethylxanthine, elixophyllin, theophyllin, theolair, nuelin, theophylline anhydrous, respbid, theocin, theo-dur
InChI Key ZFXYFBGIUFBOJW-UHFFFAOYSA-N
SMILES CN1C2=C(C(=O)N(C1=O)C)NC=N2
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione
Molecular Weight (g/mol) 180.167
PubChem CID 2153
ChEBI CHEBI:28177
Formula Weight 180.2
Physical Form Powder
Chemical Name or Material Theophylline
Show More Show Less
compliance-icons
Product Identifier
  • THEOPHYLLINE 
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Theophylline
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.