missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure 2ʹ-Fluoro-iBu-G Phosphoramidite

Product Code. 30054752 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
Amber Bottle (20-400 Thread)
Bottle (28-400 Thread)
Serum Vial Bottle
Unit Size:
0.25g
1g
500mg
Quantity:
0.25 g
0.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size Quantity
30054752 Serum Vial Bottle 1g -
30054432 Amber Bottle (20-400 Thread) 0.25g 0.25 g
17927192 Bottle (28-400 Thread) 500mg 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30054752 Supplier Thermo Scientific™ Supplier No. 27160702
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2'-Fluoro-iBu-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-Fluoro protection, for use in oligonucleotide synthesis.

2'-Fluoro-iBu-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-Fluoro protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, the highly electronegative fluorine (2'-Fluoro) modification adopts a more thermodynamically stable A-form helix on hybridization to its target. Showing improved affinity and some nuclease resistance (compared to 2'-OH), the 2'-F modification has been proven useful in antisense, siRNA, and aptamer applications. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in an amber bottle.

Available with a Serum Vial, 20-400 Finish, or 28-400 Finish neck style.

  • Serum vial neck style is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.
  • 20-400 finish neck style is compatible with most Expedite synthesizers.
  • 28-400 finish neck style is compatible with most MerMade synthesizers.

Features:

  • Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-fluoroguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
  • Molecular formula: C44H53FN7O8P
  • Base: G
  • Base Protection: iBu
  • Formula weight: 857.4 g/mol
  • CAS number: 144089-97-4
  • HPLC purity: >98%
  • 31P NMR purity: ≥98%
TRUSTED_SUSTAINABILITY

Specifications

Format Serum Vial Bottle
For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type 2in-Fluoro-iBu-G CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Guanosine (G)
Reagent Features 2in-Fluoro modified
Shipping Condition Room Temperature
Purity 98%
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
Form Powder
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.