missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure 2ʹ-OMe-U Phosphoramidite

Product Code. 17977152 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Format:
Amber Bottle (20-400 Thread)
Serum Vial Bottle
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Unit Size
17977152 1 g Amber Bottle (20-400 Thread) 1g
17907162 1 g Serum Vial Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17977152 Supplier Thermo Scientific™ Supplier No. 27204410
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2'-OMe-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-O-methyl protection, for use in oligonucleotide synthesis.

2'-OMe-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in an amber bottle.

Available with a Serum Vial, 20-400 Finish, or 28-400 Finish neck style.

  • Serum vial neck style is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.
  • 20-400 finish neck style is compatible with most Expedite synthesizers.
  • 28-400 finish neck style is compatible with most MerMade synthesizers.

Features:

  • Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyl-uridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
  • Molecular formula—C40H56N7O9P
  • Base—U
  • Formula weight—760.8 g/mol
  • CAS number—110764-79-9
  • HPLC purity—≥99%; M-11 Impurity: ≤0.01%; Single critical impurity ≤0.3%
  • 31P NMR purity—>99%; Sum of non-primary peaks at 140-152 ppm ≤0.5%
TRUSTED_SUSTAINABILITY

Specifications

Format Amber Bottle (20-400 Thread)
For Use With (Equipment) Expedite Synthesizers
Product Type 2in-OMe-U CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Uridine (U)
Reagent Features 2in-OMe modified
Shipping Condition Room Temperature
Purity 99%
Quantity 1 g
Reagent Type Phosphoramidite
Grade TheraPure™
Nucleic Acid Type RNA
Product Line TheraPure
Form Powder
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.