missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ Modifying Enzymes

Product Code. 18476768 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 U, 10 μL
10 U, 100 μL
10,000 U, 250 μL
5000 U, 100 μL
Concentration:
0.1 U/μL
1 U/μL
40 U/μL
50 U/μL
Product Type:
DNase I, RNase-free
Pyrophosphatase, Inorganic
RNase Inhibitor
Unit Size:
100µL
10µL
250µL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration Product Type unitSize
18476768 10 U, 100 μL 0.1 U/μL Pyrophosphatase, Inorganic 100µL
18446798 5000 U, 100 μL 50 U/μL DNase I, RNase-free 100µL
18496768 10,000 U, 250 μL 40 U/μL RNase Inhibitor 250µL
18416728 10 U, 10 μL 1 U/μL Pyrophosphatase, Inorganic 10µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18476768 Supplier Thermo Scientific™ Supplier No. EF022T1
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.

The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.

Features

  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product

The exceptional quality of TheraPure products is supported by:

  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1 U/μL
For Use With (Application) In vitro Transcription
Validated Application RNA synthesis
Enzyme Pyrophosphatase, inorganic
Sensitive to Heat Inactivation No
Quantity 10 U, 100 μL
Product Type Pyrophosphatase, Inorganic
Research Category Drug Development
Molecular Weight (g/mol) 32 kDa
Product Line TheraPure
Inhibitors Imidodiphosphate, α ω-glycol disphosphates, methanedial diphosphate, 1,2-ethanedial diphosphate
Form Liquid
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.