missing translation for 'onlineSavingsMsg'
Learn More

TheraPure™ Ac-rC Phosphoramidite, serum vial bottle

Product Code. 17934332 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17934332 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17934332 missing translation for 'mfr' Thermo Scientific missing translation for 'supplierNo' 27180502
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufacture oligonucleotide compounds that require controlled impurity profiles with this phosphoramidite.

TheraPure phosphoramidites are designed for oligonucleotide manufacturers requiring highly defined impurity profiles and strictly controlled processes.

Highlights

  • TheraPure PAC rA Phosphoramidites are ideal for the large scale manufacture of oligonucleotide compounds that require controlled impurity profiles in amidite manufacturing processes
  • Rigorously controlled manufacturing processes minimize levels of reactive P(III) impurities which could be incorporated into the oligonucleotide product and/or result in the accumulation of by-products which can be difficult to remove during purification
  • NMR analysis is conducted on each lot to assure that the levels of critical impurities are within specifications
  • Limited residual water improves amidite stability, eliminating the interference with oligonucleotide synthesis that can be caused by excessive water

Process and Supply Chain Control

  • A controlled raw material supply chain and established vendor partnerships ensure consistency and security of supply
  • Processes are strictly controlled and monitored from material procurement through final QC and product release under ISO9001 standards, providing batch-to-batch reproducibility
  • All TheraPure RNA phosphoramidites are supplied with lot-specific quality control data including HPLC and phosphorous NMR analyses

Product Configurations

  • TheraPure phosphoramidites are available with 2'-TBDMS protection or the nuclease resistant 2'-OMe modification
  • Fast or standard deprotection options are available
  • 2'-TBDMS RNA amidite chemistry is the widely accepted standard for high quality, large-scale synthesis of RNA oligonucleotides
TRUSTED_SUSTAINABILITY

Specifications

Format Powder
For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type Ac-rC CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Cytidine (C)
Reagent Features Fast deprotection
Shipping Condition Room Temperature
Purity 98%
Quantity 1 g
Reagent Type Phosphoramidite
Grade TheraPure™
Nucleic Acid Type RNA
Product Line TheraPure
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.