missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TheraPure phosphoramidites are designed for oligonucleotide manufacturers requiring highly defined impurity profiles and strictly controlled processes.
Highlights
- TheraPure PAC rA Phosphoramidites are ideal for the large scale manufacture of oligonucleotide compounds that require controlled impurity profiles in amidite manufacturing processes
- Rigorously controlled manufacturing processes minimize levels of reactive P(III) impurities which could be incorporated into the oligonucleotide product and/or result in the accumulation of by-products which can be difficult to remove during purification
- NMR analysis is conducted on each lot to assure that the levels of critical impurities are within specifications
- Limited residual water improves amidite stability, eliminating the interference with oligonucleotide synthesis that can be caused by excessive water
Process and Supply Chain Control
- A controlled raw material supply chain and established vendor partnerships ensure consistency and security of supply
- Processes are strictly controlled and monitored from material procurement through final QC and product release under ISO9001 standards, providing batch-to-batch reproducibility
- All TheraPure RNA phosphoramidites are supplied with lot-specific quality control data including HPLC and phosphorous NMR analyses
Product Configurations
- TheraPure phosphoramidites are available with 2'-TBDMS protection or the nuclease resistant 2'-OMe modification
- Fast or standard deprotection options are available
- 2'-TBDMS RNA amidite chemistry is the widely accepted standard for high quality, large-scale synthesis of RNA oligonucleotides
Specifications
Specifications
| Format | Powder |
| For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
| Product Type | rU CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Uridine (U) |
| Shipping Condition | Room Temperature |
| Purity | 99% |
| Reagent Type | Phosphoramidite |
| Grade | TheraPure™ |
| Nucleic Acid Type | RNA |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?