missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Thermal Cycler Power Cord (United Kingdom)

Product Code. 16865920
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16865920 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16865920 Supplier Applied Biosystems™ Supplier No. ED000650
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with the GeneAmp™ PCR System 9700, Veriti™ thermal cyclers, and 2720 Thermal Cycler

  • This power cord is appropriate for use with the GeneAmp™ PCR System 9700, Veriti™ thermal cyclers, and 2720 Thermal Cycler.
  • This version is for use in the United Kingdom.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature
For Use With (Equipment) GeneAmp 9700, Veriti Thermal Cycler, 2720 Thermal Cycler
Product Line GeneAmp, Veriti
Product Type Power Cord
Shipping Condition Room Temperature
Quantity 1 each

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.