Learn More
Manutan Thermal latex grip gloves, Matrix Hi Viz, size seven
Product Code. 12848029
Encompass_Preferred
Product Code. 12848029
Brand: Manutan A351235
Only null left
Add to basket
Add to basket
Seamless thermal high visibility liner with crinkle latex coating. Ideal for general labouring, manual handling, construction, warehousing and refuse collection.
  • Fleece lining keeps hands warm in cold conditions
  • Crinkle latex coating for excellent wet and dry grip
  • EN 388
  • Size 7