missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tubes, 0.2 mL, flat cap, yellow

Product Code. 10203173
Change view
Click to view available options
Quantity:
1000 tubes
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10203173 1000 tubes Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10203173 Supplier Thermo Scientific™ Supplier No. AB0620Y
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
Estimated Shipment: 23-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

0.2 mL individual tubes with attached flat or dome snap shut caps, available in a variety of colors

0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.

The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed
•
Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage

Hazardous:No
>Storage Condition:Ambient

PCR; Sample storage

Certifications

Certified free from human DNA, RNases, and DNases

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color Yellow
Volume (Metric) Working 0.2 mL
Volume (Metric) 0.25 mL
Quantity 1000 tubes
For Use With (Equipment) Thermal Cyclers
Material Polypropylene
Reaction Speed Standard
Closure Type Flat Cap
Format 1,000 Tubes
Product Type Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.