Learn More
Thermo Scientific™ Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Product Code. 11826340 Shop All Thermo Scientific Products
Click to view available options
Capacity:
1 x 16 mL Tubes
24 x 0.2 mL PCR Tubes
24 x 0.25/0.4 mL Microliter Tubes
24 x 0.5/0.6 mL Microliter Tubes
For Use With (Equipment):
24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Stratus Centrifuge FA Rotor
Unit Size:
Pack of 2
Pack of 24
Product Code. 11826340
Brand: Thermo Scientific™ 76003251
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor.
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor. Select from a number of Microliter Tube Adapters and PCR adapters for the Thermo Scientific™ 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Microtube Adapter
Tube Type Microtube
Capacity 24 x 0.25/0.4 mL Microliter Tubes
For Use With (Equipment) 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Tube Size 0.4/0.25 mL