Learn More
Thermo Scientific™ Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Product Code. 11836340 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
1 x 16 mL Tubes
24 x 0.2 mL PCR Tubes
24 x 0.25/0.4 mL Microliter Tubes
24 x 0.5/0.6 mL Microliter Tubes
For Use With (Equipment):
24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Stratus Centrifuge FA Rotor
Unit Size:
Pack of 2
Pack of 24
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity For Use With (Equipment) Unit Size
11836340 24 x 0.5/0.6 mL Microliter Tubes 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid Pack of 24
10751313 1 x 16 mL Tubes Stratus Centrifuge FA Rotor Pack of 2
11846340 24 x 0.2 mL PCR Tubes 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid Pack of 24
11826340 24 x 0.25/0.4 mL Microliter Tubes 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11836340 Supplier Thermo Scientific™ Supplier No. 76003252
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor.
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor. Select from a number of Microliter Tube Adapters and PCR adapters for the Thermo Scientific™ 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Microtube Adapter
Tube Type Microtube
Capacity 24 x 0.5/0.6 mL Microliter Tubes
For Use With (Equipment) 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Max. RCF 21100
Max. Speed 14800
Tube Size 0.6/0.5 mL