Learn More
Thermo Scientific™ Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Shop All Thermo Scientific ProductsDescrizione
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor. Select from a number of Microliter Tube Adapters and PCR adapters for the Thermo Scientific™ 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifica
Specifica
| Type | PCR Tube Adapter |
| Tube Type | PCR |
| Capacity | 24 x 0.2 mL PCR Tubes |
| For Use With (Equipment) | 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid |
| Tube Size | 0.2 mL |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.