missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Filtered Pipette Tips with Breakthru Economy Packaging

Product Code. 10497341
Change view
Click to view available options
Volume (Metric):
0.1 to 20 μL
Sterility:
Sterile
Filtered:
Filtered
Unit Size:
Pack of 960
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Unit Size
10497341 0.1 to 20 μL Sterile Filtered Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10497341 Supplier Thermo Scientific™ Supplier No. 2149P50
Spotlight Offers
Use promo code 28037
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • The stackable Breakthru™ packaging uses 85% less packaging materials and 53% less space
  • Breakthru packaging is universal to all 9mm trays and works with Art™ self-sealing barrier tips Tips are racked
  • ART 20P and Pure 200 are sterile; ART 200 are nonsterile
TRUSTED_SUSTAINABILITY

Specifications

Description ART 20P
Volume (Metric) 0.1 to 20 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Product Line ART
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.