Learn More
Thermo Scientific™ Barnstead™ Water Purification System Replacement Cartridges
Product Code. 13204749 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cartridge
Organicfree Cartridge
Pretreatment Cartridge
TFM Membrane
For Use With (Equipment):
Easypure Reverse Osmosis System
Nanopure Infinity System
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
13204749 Pretreatment Cartridge Nanopure Infinity System Each
11810810 Cartridge Nanopure Infinity System Each
11820810 Organicfree Cartridge Nanopure Infinity System Each
11810920 TFM Membrane Easypure Reverse Osmosis System Each
10080614 Pretreatment Cartridge Easypure Reverse Osmosis System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13204749 Supplier Thermo Scientific™ Supplier No. D50251
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
For Easypure™ RO water purification system
TRUSTED_SUSTAINABILITY
Especificaciones
Feedwater Source Deionized, Tap
Resin Type Activated Carbon and Macroreticular
For Use With (Equipment) Nanopure Infinity System
Type Pretreatment Cartridge
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.