missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BioBasic™ AX Javelin™ Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Brand: Thermo Scientific™ 73105-014006
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- Weak anion exchange phase for multiple charged species
- 300Å pore size for enhanced protein and peptide separations
- Suitable for HILIC retention and separation of highly polar molecules
- Superb stability under demanding pH conditions
Specifications
5 μm | |
4 mm | |
BioBasic |
10 mm | |
AX |