missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CpG Methylated Jurkat Genomic DNA

Product Code. 11931421 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
15 μg
Unit Size:
15µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11931421 15 μg 15µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11931421 Supplier Thermo Scientific™ Supplier No. SD1121
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Methylated human male genomic DNA from Jurkat cells used as a control in DNA methylation analysis.

Genomic DNA was isolated from Jurkat E6.1 (human acute T-cell leukemia, clone E6.1) cells grown to confluency in RPMI media supplemented with 10% bovine fetal serum. Isolated genomic DNA was treated with CpG Methyltransferase, repurified and dissolved in TE buffer. ≥98% CpG methylation level confirmed by bisulfite sequencing.

Thermo Scientific CpG Methylated Jurkat Genomic DNA is human male genomic DNA from Jurkat cells, enzymatically methylated with CpG Methyltransferase (M.SssI). Genomic DNA was isolated from Jurkat (human acute T-cell leukemia) cells grown to confluency in RPMI media supplemented with 10% bovine fetal serum. Isolated genomic DNA was treated with CpG Methyltransferase, repurified, and dissolved in TE buffer. ≥98% CpG methylation level confirmed by bisulfite sequencing.

Highlights:
• Pure - DNA free from inhibitors, proteins, and RNA
• Convenient - ready to use 100 ng/μL concentration
• Compatible with downstream applications
• Complete methylation of cytosines in CpG dinucleotides

Applications
As controls in the following:
• Bisulfite sequencing
• Combined Bisulfite Restriction Analysis (COBRA)
• Methylation-sensitive Single-Nucleotide Primer Extension (Ms-SNuPE)
• A260/230 ratio: ≥ 2
• A260/280 ratio:1.75 to 1.98

Quality Control
• Predominant > 30 kb band is observed during agarose gel electrophoresis. DNA concentration is determined spectrophotometrically. The absence of nucleases is confirmed by labeled oligonucleotide tests.
• CpG methylated DNA was protected from digestion by the methylation sensitive restriction enzyme HpaII as determined by qPCR analysis. Following qPCR, equivalent Cq values for HpaII digested and control undigested DNA are observed.
• DNA purity is confirmed through a functional qPCR test in which Cq values of MspI and HpaII digested template DNA must differ by ≥ 5 cycles.
• The absence of mycoplasma DNA is confirmed by PCR.
• As controls in bisulfite sequencing, Combined Bisulfite Restriction Analysis (COBRA), Methylation-sensitive Single-Nucleotide Primer Extension (Ms-SNuPE)

Order Info

Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 ng/μL
Format Tube
For Use With (Application) Sequencing
Includes [other]
Product Type CpG Methylated Jurkat Genomic DNA
Quantity 15 μg
Species Human
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.