Learn More
Thermo Scientific™ GeneJET NGS Cleanup Kit
Thermo Scientific GeneJET NGS Cleanup Kit provides protocols optimized specially for fast DNA fragment cleanup or adapter removal procedures.
Brand: Thermo Scientific™ K0852
497.00 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : NEW
Additional Details : Weight : 2.01000kg
Description
Thermo Scientific GeneJET NGS Cleanup Kit provides protocols optimized specially for fast DNA fragment cleanup or adapter removal procedures. The clean up protocol results in higher than 70% recovery of DNA fragments larger than 150 bp and more than 97% removal of adapters. Adapter removal protocol results in more than 70% recovery of DNA fragments larger than 200 bp and more than 99% removal of adapters. The entire procedure takes 5 minutes for DNA fragment library cleanup from enzymatic reaction mixtures and 7 minutes for adapter removal protocol. Spin columns technology in combination with unique binding buffer allows to get the reproducible yields of DNA fragments in the wide starting material range (5 ng - 1 μg of DNA fragments).
The GeneJET NGS Cleanup Kit is compatible with Illumina™, Life Technologies library preparation workflows.
Highlights
• Fast—high quality DNA fragments obtained within 5 min
• Robust—reliable cleanup and size selection from different reaction mixtures and buffers, such as end-conversion, ligation, adapter addition, PCR mixtures, fragmentation buffers
• Efficient—DNA recovery up to 90%
• NGS-grade purity—free of primers, adapters, nucleotides, enzymes and other reaction components inhibiting NGS workflow.
Applications
• Purified DNA fragments are ideal for use in:
• PCR and qPCR
• Next Generation Sequencing workflow
Includes
• Binding Buffer (Cat. No. R2811)
• Prewash Buffer (conc.) (Cat. No. R2821)
• Wash Buffer (conc.)(Cat. No. R2831)
• Elution Buffer (10 mM Tris-HCl, pH 8.5)
• DNA Purification Micro Column & Collection Tube
• Collection Tube
Specifications
Spin Column | |
DNA | |
Not High-throughput Compatible (Manual) | |
GeneJET | |
DNA from Enzymatic Reactions (e.g. PCR) |
Spin Column | |
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), PCR | |
250 Preps | |
250 Preps | |
5 min. |
For Research Use Only. Not for use in diagnostic procedures.