missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Luminaris HiGreen qPCR Master Mix, fluorescein

Product Code. 13545260 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
1250 Reactions
250 Reactions
500 Reactions
5000 Reactions
Unit Size:
1250 reactions
250 reactions
500 reactions
5000 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
13545260 250 Reactions 250 reactions
13565260 5000 Reactions 5000 reactions
13535260 500 Reactions 500 reactions
13555260 1250 Reactions 1250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13545260 Supplier Thermo Scientific™ Supplier No. K0981
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Luminaris HiGreen Fluorescein qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer. The master mix contains fluorescein passive reference dye for specific real-time platfo

Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.

  • UDG in the master mix to prevent carry-over contamination
  • Equal performance of blue and colorless master mixes
  • Versions with premixed ROX or fluorescein for different real-time platforms
  • Standard cycling protocols
  • Excellent qPCR performance
  • Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity: single copy detection
  • Wide linear range: accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience: ready-to-use 2X master mix
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 2 x 1.25 mL Luminaris HiGreen Fluorescein qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 250 x 20 μL reactions. The master mix contains fluorescein passive reference dye for specific real-time platforms.
• 2 x 1.25 mL Nuclease-Free Water

Store at -20°C.
Detection Method SYBR
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5
Product Line Luminaris
Product Type qPCR Master Mix
Sufficient For 250 Reactions
For Use With (Application) Gene Expression
Fidelity (vs. Taq) 2 X
No. of Reactions 250 Reactions
Sample Type DNA, dsDNA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.