missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagJET Plasmid DNA Kit

Product Code. 13565180 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 x 96 Preps
96 Preps
Unit Size:
96 tests
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13565180 96 Preps 96 tests
13585180 4 x 96 Preps Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13565180 Supplier Thermo Scientific™ Supplier No. K2791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.

Thermo Scientific MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.

The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET Magnetic Beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification for low sample throughput users.

The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors. Purified plasmid DNA can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.

Highlights
High yields—up to 5 μg of plasmid DNA from 1 mL of bacterial culture
Pure—isolated DNA is immediately ready-to-use in downstream application such as sequencing, transcription, digestion with restriction enzymes or transfection of robust cell lines
Flexible—automation and manual processing protocols available
Repeatable results and robust performance

Applications
Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
• FastDigest or conventional restriction digestion
• Automated fluorescent and radioactive sequencing
• PCR
• In vitro transcription
• Transformation
• Transfection of robust cell lines

TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Sample Type Bacteria
For Use With (Application) Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual)
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line MagJET
Product Type Plasmid DNA Kit
Quantity 96 Preps
Target Plasmid DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.