missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MuSeek Library Preparation Kit, Illumina compatible

Product Code. 15206187 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
12 Reactions
Unit Size:
12 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15206187 12 Reactions 12 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15206187 Supplier Thermo Scientific™ Supplier No. K1361

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Illumina-compatible next-gen sequencing (NGS) libraries from as little as 50ng of starting DNA. DNA fragmentation and transposon-tagging are accomplished in one step.

Thermo Scientific MuSeek Library Preparation Kit, Illumina compatible is designed for generation of high-quality genomic DNA libraries for sequencing with the Illumina™ MiSeq™ and HiSeq™ systems. The fast protocol utilizes the MuA transposase enzyme, which catalyzes simultaneous fragmentation of double-stranded target DNA and tagging the fragment ends with transposon DNA. In a subsequent PCR step the platform-specific adapters are added using a robust and accurate Thermo Scientific™ Phusion™ High-Fidelity DNA polymerase. Starting with only 50ng or 100ng of sample the protocol can be used to generate 100 bp to 1,000 bp insert libraries. The kit contains components sufficient for 12 fragmentation reactions and the subsequent adapter-addition polymerase chain reactions. Sequencing primers required for sequencing libraries prepared by this method are provided as well.

Highlights

  • Convenient – no physical shearing, end-repair and ligation steps
  • Fast – generates NGS fragment libraries in less than 90 minutes
  • Low sample input – DNA fragment libraries from 50ng DNA input
  • Indexing compatible – MuSeek Index Set 1 enables preparation of up to 24 dual-indexed DNA libraries for sequencing in one run
Applications
  • DNA fragment library preparation for downstream use in next generation sequencing workflow
  • Library preparation from gDNA, ds DNA, bacterial DNA, DNA amplicons
Includes
  • MuSeek Enzyme Mix, IL
  • MuSeek Fragmentation Reaction Buffer
  • MuSeek Stop Solution
  • Water, nuclease-free
  • Control DNA
  • MuSeek Adapter Addition Primer Mix, IL
  • MuSeek Adapter Addition Reaction Buffer, IL
  • Phusion™ Hot Start II High-Fidelity DNA Polymerase, IL
  • MuSeek Sequencing Primer, Read 1
  • MuSeek Sequencing Primer, Read 2
  • MuSeek Sequencing Primer, Index Read
Notes
The kit is shipped on dry ice. Upon arrival store at -20°C, avoid repeated freeze-thaw cycles. The Stop Solution can be stored at room temperature.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Library Preparation Kit
Libraries Fragment Library
For Use With (Equipment) Illumina™ HiSeq 2500, Illumina ™ MiSeq
Sample Type DNA
Sequencing Type Genome & DNA Sequencing
Workflow Step Library Generation
Product Line MuSeek
For Use With (Application) NGS library preparation
Quantity 12 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.