missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Animal Tissue Direct PCR Kit (without sampling tools)

Product Code. 18857343 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
18857343 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 18857343 Supplier Thermo Scientific™ Supplier No. F140WH
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Thermo Scientific Phire Animal Tissue Direct PCR Kit has been developed for amplification of DNA directly from a wide variety of animal tissues including mice, fish, birds and insects.

Thermo Scientific Phire Animal Tissue Direct PCR Kit has been developed for amplification of DNA directly from a wide variety of animal tissues including mice, fish, birds, and insects. The kit contains all the necessary components for amplification of DNA directly from animal tissues: optimized PCR reagents, dilution buffer, and DNARelease additive, which can be used to improve the release of DNA from animal tissues. In addition, the kit includes control primers for positive control reactions that are universal and work with numerous animal species.

Features

  • No need for time-consuming and expensive DNA purification steps
  • Very little sample material required
  • Two simple protocols for various applications
  • Phire Hot Start II DNA Polymerase provides high yields of specific product with short extension time (20 s/kb).

Notes

  • A detailed manual is provided that describes protocols when working with different types of tissues.
  • The optimal annealing temperature for Phire DNA Polymerase may differ significantly from that of Taq-based polymerases.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Phire Hot Start II DNA Polymerase, 200 μL
• 2X Phire Animal Tissue PCR Buffer, 5 x 1.25 mL
• Control Primer Mix (25 μM each), 40 μL
• Dilution Buffer, 2 x 5 mL
• DNARelease Additive, 3 x 100 μL
• Gel Loading Dye, 3 x 1 mL

Store at -20°C.
GC-Rich PCR Performance Medium
PCR Method Direct PCR
Polymerase Phire Hot Start II DNA Polymerase
Reaction Speed Fast
Product Type Direct PCR Kit
Quantity 500 reactions
Shipping Condition Wet or Dry Ice
Hot Start Built-In Hot Start
No. of Reactions 500 reactions
Overhang Blunt
Reaction Format Separate Components
Sample Type Tissue
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.