missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion U Hot Start PCR Master Mix

Product Code. 15205146 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
15205146 500 Reactions 500 reactions
15295136 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 15205146 Supplier Thermo Scientific™ Supplier No. F533L
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology.

Thermo Scientific Phusion U Hot Start PCR Master Mix is a convenient 2X formula that contains Phusion U Hot Start DNA Polymerase, nucleotides, and an optimized reaction buffer including MgCl2.

Phusion U DNA Polymerase is a novel engineered high-fidelity enzyme developed using fusion technology. Designed with a proprietary mutation in the dUTP binding pocket of Phusion polymerase, Phusion U polymerase is able to overcome important limitations of proofreading enzymes to incorporate dUTP and read through uracil present in DNA templates.

Phusion U polymerase features properties of Phusion DNA polymerases and offers the ability to amplify long amplicons up to 20 kb and a high specificity with affibody based hot start.

These features make Phusion U Hot Start DNA Polymerase an excellent choice for such applications as amplification of bisulfite-converted or damaged DNA as well as use of carryover contamination control.

Highlights

  • High-fidelity DNA polymerase (25X Taq)
  • Engineered to incorporate dUTP and amplify uracil containing templates
  • Hot start for reduced nonspecific amplification and primer degradation
  • Only template and primers need to be added for use
  • Short extension times (15–30 s/kb)

Applications

  • Amplification of bisulfite-converted DNA
  • Amplification of damaged or aged DNA
  • Carry-over contamination control
  • Uracil-excision based (USER) cloning methods

Using Phusion DNA Polymerases

  • Annealing rules for Phusion DNA polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • Phusion U Hot Start DNA Polymerase
• 2X reaction buffer
• 3 mM MgCl2
• 400 μM each of dATP, dTTP, dCTP and dGTP

Master mix is supplied with nuclease-free water.
Store at -20°C.
GC-Rich PCR Performance Medium
Polymerase Phusion U Hot Start DNA Polymerase
Reaction Speed Fast
Product Type Hot Start PCR Master Mix
Quantity 500 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 25X
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 20 kb or less
Show More Show Less
Do Phusion DNA Polymerases add the non-template dependent 3'-A overhang?

Phusion DNA Polymerases generate blunt end products; therefore, blunt end cloning is recommended. If TA cloning is required, it can be performed by adding A overhangs to the blunt PCR product with e.g. Taq DNA Polymerase (Cat. No. EP0401). However, before adding the overhangs it is very important to remove all the Phusion DNA Polymerase by purifying the PCR product carefully, as the proofreading activity in Phusion DNA Polymerase is very strong. Any remaining Phusion DNA Polymerase will degrade the A overhangs, thus creating blunt ends again.

Can Phusion DNA Polymerases extend at 1 second/kb?

Yes it is possible, especially when amplifying smaller amplicons. Processivity of Phusion DNA Polymerases is 10 times that of Pfu. We recommend extension times of 15 s/kb for Phusion Flash PCR Master Mix. 15 s/kb is a conservative value that we can promise to work with almost any amplicon. In many cases, significantly shorter extension times (0-5 s/kb) can be used without compromising the yield. What separates Phusion Flash DNA Polymerase from other fast polymerases is that all steps in the PCR protocol can be shortened, including annealing and denaturation. This results in extremely fast protocols as compared with any other polymerase.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.