Learn More
Thermo Scientific™ Forma™ Chamber Cooling Coil
Product Code. 10145382 Shop All Thermo Scientific Products
Change view
Click to view available options
CO2 Sensor Technology:
TC Sensor or IR Sensor
Unit Size:
Each
Product Code. CO2 Sensor Technology unitSize
10145382 TC Sensor or IR Sensor Each
1 options
Product Code. 10145382 Supplier Thermo Scientific™ Supplier No. 190645
  • ONLINE EXCLUSIVE PRICE
    1670.00 €
    1570.00 € / Pack of 1
    Save 100.00 €
    5% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
For Use With: Forma Series II Water jacketed CO2 Incubator
Precisely control the CO2 and temperature with Thermo Scientific™ Forma™ Series CO2 Incubators. Forma™ CO2 Incubator Accessories are important components for optimal operation.
  • Precise control
  • Choice of TC or IR sensors
  • Superior parameter recovery characteristics
  • Continuous contamination control technology
TRUSTED_SUSTAINABILITY
Specifications
CO2 Sensor Technology TC Sensor or IR Sensor
For Use With (Equipment) Forma Series II Water jacketed CO2 incubator
Lighting LED
Type Chamber Cooling Coil