Learn More
Thermo Scientific™ High Purity Two Stage Brass Gas Regulators
Product Code. 10544315 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
CGA 350 with Combi Filter
CGA 350 with Triple Filter
CGA 580 with Triple Filter
CGA 590 with Combi Filter
DIN 1 with Triple Filter
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10544315 CGA 580 with Triple Filter Each
10448854 DIN 1 with Triple Filter Each
10046844 CGA 350 with Triple Filter Each
10065674 CGA 350 with Combi Filter Each
10117984 CGA 590 with Combi Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10544315 Supplier Thermo Scientific™ Supplier No. 60180902
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Recommended for non-corrosive analytical and process applications requiring precise, stable, delivery pressure control
  • For non-corrosive analytical and process applications
  • Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
  • Specially designed, convoluted, stainless steel diaphragm provides goodregulating performance and maximum purity integrity without the needfor a soft seal
  • Capable of withstanding an internal vacuum
  • All regulators supplied with 1/8 in. stainless steel output adaptor
  • Built in check valve allows for cylinder change without contaminating the gas line
TRUSTED_SUSTAINABILITY
Specifications
Range 0 to 200 psig
Type CGA 580 with Triple Filter