Learn More
Thermo Scientific™ Convenience Receptacle
Shop All Thermo Scientific Products
Click to view available options
Type:
Convenience Receptacle
Unit Size:
Each
Description
Provide electricity to an electrical device inside your environmental chamber or incubator.
Specifications
Specifications
| Description | RS-485 interface (compatible with 1535 Monitor/Alarm only) Data Output |
| For Use With (Equipment) | 3962 Environmental Chamber/Incubator |
| Lighting | LED |
| Type | Convenience Receptacle |
| Installation | Factory installed |