Learn More
Thermo Scientific™ Perforated Shelf Kit
Shop All Thermo Scientific Products
Click to view available options
Type:
Shelf
Unit Size:
Each
Description
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.
Specifications
Specifications
| Description | Perforated Stainless Steel Shelf and Channels |
| For Use With (Equipment) | Large Capacity Reach-In CO2 Incubator |
| Lighting | LED |
| Material | Stainless Steel |
| Type | Shelf |
| Includes | One Shelf, with channels |
| Quantity | 1 shelf |