Learn More
Thermo Scientific™ Adjustable Reinforced Shelf with Channels
Product Code. 15523771 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Shelf
Unit Size:
Each
Product Code. Type Unit Size
15523771 Shelf Each
1 options
Product Code. 15523771 Supplier Thermo Scientific™ Supplier No. 224161
  • ONLINE EXCLUSIVE PRICE
    638.18 €
    573.00 € / Pack of 1
    Save 65.18 €
    10% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Material: Stainless Steel; For Use With: Large capacity reach-in CO2 Incubator; ; Environmental chambers 3948, 3907 and 3962
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.
TRUSTED_SUSTAINABILITY
Specifications
Description Reinforced Stainless Steel Shelf and Channels (max. load 68kg)
For Use With (Equipment) Large Capacity Reach-In CO2 Incubator
Lighting LED
Material Stainless Steel
Type Shelf