missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Mini Gel Incubation Trays are a convenient way to process mini-sized protein gels and Western blot membranes for use in general immunostaining applications. The trays are reusable, optically clear, and supplied with a sealable lid.
Specifications
Specifications
| Formulation | Clear, semi-rigid plastic (APET, 0.6mm thick) trays (95mm x 95mm base) and snap-on lids |
| Quantity | 10 Each |
| Sufficient For | Incubating mini-sized protein electrophoresis gels |
| Product Type | Mini Gel Incubation Tray |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?