missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MuSeek Index Set 1, Illumina compatible

Product Code. 15216187 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
24 Dual Indices (4 x 6)
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15216187 24 Dual Indices (4 x 6) Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15216187 Supplier Thermo Scientific™ Supplier No. K1551

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enables pooling of up to 24 dual-indexed libraries in one sequencing run

MuSeek Index Set 1, Illumina compatible, is designed for generation of indexed genomic DNA libraries for sequencing on the Illumina MiSeq™ and HiSeq™ instruments. The set provides 4 indexed P5 adapters and 6 MuSeek indexed P7 adapters. This enables pooling of up to 24 dual-indexed libraries and conducting multiplexed sequencing analysis. The MuSeek Index Set 1, in combination with MuSeek Library Preparation kit, Illumina compatible, allows fast and simple indexed library construction from 50ng of sample DNA, eliminating the need for physical shearing, end conversion and adapter ligation steps.

Highlights

  • Enables sequencing of up to 24 dual-indexed fragment libraries in a single sequencing run.
Applications
  • Indexed DNA fragment library preparation for next generation sequencing.
Includes
  • MuSeek Indices M501-4
  • MuSeek Indices M701-6

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol).
Includes:
o Tango™ CMKLR1-bla U2OS DA cells
o LiveBLAzer™-FRET B/G Loading Kit, 70μg
o Solution D
o Tango™ CMKLR1-bla U2OS Assay Protocol
o LiveBLAzer™-FRET B/G Loading Protocol
o Certificate of Analysis
The Tango™ CMKLR1-bla U2OS DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Format Kit
Sample Type DNA
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) Illumina™ HiSeq 2500, Illumina ™ MiSeq
For Use With (Application) Sequencing
Libraries Fragment Library
Product Line MuSeek
Product Type MuSeek Index Set 1
Quantity 24 Dual Indices (4 x 6)
Workflow Step Library Generation
Form Solution
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.