Learn More
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles
GREENER_CHOICE
Product Code. p-4435289
Click to view available options
Neck Finish:
24-400
28-400
Unit Size:
Pack of 25
Product Code. 10385651
Brand: Thermo Scientific™ 7124010002
Only null left
Add to basket
Add to basket
Through-the-top tubing with closure and stem molded in one piece
  • Closure and stem molded in one piece for leakproof service
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Leakproof†
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Neck Finish 24-400
For Use With (Equipment) Wash bottle