missing translation for 'onlineSavingsMsg'
Learn More

Nunc™ Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes
GREENER_CHOICE

Product Code. 15533694
Change view
Click to view available options
Holds:
2 mL Cryogenic Tubes
5 mL Cryogenic Tubes
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15533694 2 mL Cryogenic Tubes Pack of 10
15543694 5 mL Cryogenic Tubes Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15533694 Supplier Nunc™ Supplier No. 331825BR
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes

Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes.

  • Non-sterile
  • Case of 10
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) For 2mL Cryobank, non-sterile
Holds 2 mL Cryogenic Tubes
Quantity 10/pk., 10/cs.
Type Cryogenic Tube Storage Rack (w/Latch)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.