Learn More
Thermo Scientific™ Owl™ ADJ2 Adjustable System Accessories
Description
- Extremely versatile adjustable system.
- The height of the upper buffer chamber (UBC) may be adjusted from 20 to 70cm by sliding the UBC up or down the two support poles.
Specifications
Specifications
| For Use With (Equipment) | ADJ1 Giraffe™ Adjustable Gel Electrophoresis System |
| Type | Gasket |