missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Poly HRP Dilution Buffer

Product Code. 10495325 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10495325 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10495325 Supplier Thermo Scientific™ Supplier No. N501
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifically designed to optimize performance of Poly-HRP Streptavidin

Thermo Scientific Pierce Poly-HRP Dilution Buffer is optimized to ensure optimal performance with Poly-HRP Streptavidin.

Features of Poly-HRP Dilution Buffer:

• Formulation: 1% biotin-free casein in a PBS buffer solution.
• Application: Dilution of Poly-HRP Streptavidin (Product # N200): Gently invert bottle and swirl to mix before use. Do not vortex. Use at full strength. Refer to Poly-HRP Streptavidin Certificate of Analysis for recommended dilution ranges. Dilute Poly-HRP Streptavidin immediately before use.

Related Products
Poly-HRP Streptavidin

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dilution Buffer
Physical Form Liquid
Quantity 1 L

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.