Learn More
Thermo Scientific™ Rheodyne 9010 Sample Injector
Shop All Thermo Scientific Products
Click to view available options
Material:
PEEK
Unit Size:
Each
Description
Includes
20μL sample loop
Single-mode sample injector designed for the complete filling method.
Includes: 20μL sample loop
- Compatible with sample loop sizes 5μL to 10mL
- PEEK stator
- Position sensing switch provides a reproducible start signal
Specifications
Specifications
| Volume (Metric) Sample | 20 μL |
| For Use With (Equipment) | Sample Loops 5 to 50 μL |
| Material | PEEK |
| Type | Sample Injector |
| No. of Ports | 6 Ports |
| Stator Material | PEEK |