Learn More
Thermo Scientific™ Round Buckets for 4-Place Swinging Bucket Rotor
Product Code. 10210362 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Swinging bucket rotor
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10210362 Swinging bucket rotor Each
1 options
Product Code. 10210362 Supplier Thermo Scientific™ Supplier No. 75007555
  • ONLINE EXCLUSIVE PRICE
    326.63 €
    297.00 € / Pack of 1
    Save 29.63 €
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
4 x 100mL capacity buckets fit the Thermo Scientific Swinging 4-place rotor (75-007-591)
Order Info
Sold individually; order correct quantity to equip rotor. Biocontainment lids (75-007-598) sold separately. Optional adapters accept assorted tube styles and volumes from 1.5 to 50mL. No adapter is required for 100mL bottle.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Swinging bucket rotor