missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Histocentre™ 3 Embedding Center Accessories

Product Code. 12794846 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Forcep
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12794846 Forcep Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12794846 Supplier Epredia™ Supplier No. B64110040
Quantity Price
1 905.00 € / Pack of 1
4 856.22 € / Pack of 1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Red
For Use With (Equipment) Histocentre 3 Embedding Center
Product Type Forcep
Quantity Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.