Learn More
Thermo Scientific™ Sorvall™ HT6 Tube Adapters for 6-Place Swinging Bucket Rotor with Round Buckets
Product Code. 12468682 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Swinging bucket rotor for HT6 centrifuge
Unit Size:
Pack of 6
Product Code. For Use With (Equipment) Unit Size
12468682 Swinging bucket rotor for HT6 centrifuge Pack of 6
1 options
Product Code. 12468682 Supplier Thermo Scientific Supplier No. 11178308
  • ONLINE EXCLUSIVE PRICE
    859.28 €
    786.00 € / Pack of 6
    Save 73.28 €
    8% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Sorvall HT6 centrifuge
Fits in rotor 11178252 when equipped with round buckets 11178135 (sealed) or 11178136 (unsealed).
Order Info
Sold in packs of 6. One pack will fill the rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Swinging bucket rotor for HT6 centrifuge