Learn More
Thermo Scientific™ Sorvall™ Microplate Carrier
Product Code. 10639142 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Sorvall swing bucket rotor
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10639142 Thermo Scientific Sorvall swing bucket rotor Each
1 options
Product Code. 10639142 Supplier Thermo Scientific™ Supplier No. 75006449
  • ONLINE EXCLUSIVE PRICE
    625.00 €
    601.00 € / Pack of 1
    Save 24.00 €
    3% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accommodates 4 x 4 standard microplates (or one deepwell plate)
  • For use with Four-Place Swinging Bucket Rotors 75-006-441 and 75-006-446
  • Maximum speed/RCF: 4150rpm/3060xG
  • Maximum carrier height: 60mm
Order Info
Sold individually; order four to fully equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Sorvall swing bucket rotor