missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance -20°C Manual Defrost Freezers
BIOPHARMA

Product Code. 16337191 Shop All Thermo Scientific Products
Change view
Click to view available options
Plug Type:
CEE 7/7
UK/Ireland or EU (CEE 7/7)
Electrical Requirements:
208-230V, 50Hz
Dimensions (D x W x H) Interior:
28.5 x 24 x 58 in.
28.5 x 30.0 x 58.0 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Electrical Requirements Dimensions (D x W x H) Interior Unit Size
16337191 UK/Ireland or EU (CEE 7/7) 208-230V, 50Hz 28.5 x 30.0 x 58.0 in. Each
15658416 CEE 7/7 208-230V, 50Hz 28.5 x 24 x 58 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 16337191 Supplier Thermo Scientific™ Supplier No. TSX3020FV
  • ONLINE EXCLUSIVE PRICE
    10190.00 €
    8025.29 € / Pack of 1
    Save 2164.71 €
    21% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers offer a sustainable choice for clinical and laboratory storage

Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers are designed are designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated. The design supports sample protection and sustainability objectives for the storage of enzymes, reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Up to 50% less energy usage than conventional refrigerant models*
  • Natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP)
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Temperature range of -35° to -15°C, factory setpoint at -20°C
  • Lower HVAC costs as a result of less heat emission into the environment
  • Easy to use microprocessor control with setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
  • Optional chart recorders available
  • Warranty varies by country, please contact us for details

*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models

TRUSTED_SUSTAINABILITY

Specifications

Type High-Performance Freezer
Capacity (English) 29.2 cu. ft.
Capacity (Metric) 827 L
Temperature Range -25°C to -15°C
Temperature (Setpoint) -25°C to -15°C (-20°C)
Door Style Solid
No. of Doors 1
Shelves 4
Certifications/Compliance UL, cUL, CE
Style Upright
Display Microprocessor
Product Line TSX Series
Defrost Manual
Model TSX3020FV
Dimensions (DxWxH) Exterior 38.9 x 34.0 x 78.5 in.
Dimensions (D x W x H) Interior 28.5 x 30.0 x 58.0 in.
Depth (English) Exterior 38.9 in.
Depth (English) Interior 28.5 in.
Depth (Metric) Exterior 99.0 cm
Depth (Metric) Interior 72.3 cm
Height (English) Exterior 78.5 in.
Height (English) Interior 58 in.
Height (Metric) Exterior 199.4 cm
Height (Metric) Interior 147.3 cm
Width (English) Exterior 34.0 in.
Width (English) Interior 30 in.
Width (Metric) Exterior 86.4 cm
Width (Metric) Interior 76.2 cm
Monitoring Options Wireless
Chart Recorder Optional
Smart View Compatibility Smart-Vue Compatible
Amperage 15 A
Plug Type UK/Ireland or EU (CEE 7/7)
Voltage 230 V
Electrical Requirements 208-230V, 50Hz
Frequency 50/60 Hz
Compressor V-drive
Refrigerant R290
Shipping Weight (English) 483 lb.
Shipping Weight (Metric) 219 kg
Weight (English) 415 lb.
Weight (Metric) 188 kg
Rack Capacity 4
Show More Show Less

*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.