missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pEF1α Tluc16-DD Vector for Luciferase Assays

Product Code. 15362637 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15362637 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15362637 Supplier Thermo Scientific™ Supplier No. 88241
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Insert promoter sequences for genes of interest into Thermo Scientific™ Tluc16 Vectors to measure gene regulation using the intracellular TurboLuc™ 16 (Tluc16) luciferase reporter in luciferase assays.

The Thermo Scientific pMCS Tluc16 Vectors are multiple cloning site (MCS) vectors designed to accept a promoter sequence for study of gene regulation using the intracellular TurboLuc™16 (Tluc16) luciferase reporter.

  • Intracellular Tluc16 luciferase gene, optimized for high expression in mammalian systems
  • Multiple cloning sites provide versatility for transfer of regulatory elements from one vector to another
  • Synthetic polyA terminator and Transcriptional Pause Site (TPS) included upstream of MCS to minimize non-specific transcriptional read-through
  • Dual-destabilization (DD) technology reduces accumulation of Tluc16 luciferase mRNA and protein in cells, enhancing the responsiveness of the assay
  • Optimized minimal core promoter (minP) and 5' UTR region for efficient expression of Tluc16 luciferase
  • Hygromycin resistance gene for use as selection marker to generate mammalian stable cell lines
  • Prepared pNF-κB, and pCRE Tluc16-DD Vectors are designed to monitor the activation of NFκB protein and cAMP-binding protein (CREB) in mammalian cells
  • Prepared pEF1α variety includes a modified version of the Elongated Factor 1α (EF1α) promoter for constitutive Tluc16 luciferase expression in mammalian cells
Tluc16 luciferase is a 16 kDa, novel, intracellular luciferase derived from the marine copedod Metridia luciferase family. The wild-type luciferase has been modified to reduce its size, increase its brightness, and enable its intracellular expression. A synthetic polyA terminator and a Transcriptional Pause Site (TPS) are included upstream of the MCS to minimize non-specific transcriptional read-through. The Tluc16 luciferase activity can be measured using the Thermo Scientific TurboLuc™ One-Step Glow Assay Kit.

TRUSTED_SUSTAINABILITY

Specifications

Type Transcriptional Reporter Vector
Cloning Method Restriction Enzyme ⁄ MCS
Delivery Type Transfection
Quantity 10 μg
Content And Storage Store in freezer (-5 to -30°C).
Reporter Gene TurboLuc™16 Luciferase
For Use With (Application) Reporter Assays
Promoter EF-1α

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.