missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Thermoblock for 24 Cryo Tubes

Product Code. 16883543 Shop All Eppendorf Products
Change view
Click to view available options
Diameter (Metric) Outside:
12.5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Unit Size
16883543 12.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16883543 Supplier Eppendorf™ Supplier No. 5318000001
  • ONLINE EXCLUSIVE PRICE
    685.00 €
    671.00 € / Pack of 1
    Save 14.00 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eppendorf SmartBlocks are available in different formats to serve the various vessel types in the lab.

  • Optimized block design supports optimal temperature transfer within your sample to receive excellent results
  • Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Every SmartBlock is individually inspected in production for temperature accuracy and is delivered including serial-number specific certificate for documentation purpose
  • Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept)
  • Automatic block recognition for safe handling
  • Eppendorf QuickRelease™ technology enables fast and simple block exchange - no need for tools
  • Limited to ThermoMixer C and ThermoStat C
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Eppendorf ThermoMixer™ C and Eppendorf ThermoStat™ C
For Use With (Application) 24 Cryo Tubes
Sample Volume (Metric) 1.5 to 2 mL
Speed 2000 rpm
Diameter (Metric) Outside 12.5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.