missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific THERMOFLUID SYNTH 60, 5L

Brand:  Thermo Scientific 9990209


THERMOFLUID SYNTH 60, 5L

Product Code. 11651091

  • ONLINE EXCLUSIVE PRICE
    274.00 €
    261.00 € / 5 L
    Save 13.00 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • bath liquid synth 60

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H413-May cause long lasting harmful effects to aquatic life.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contain: hydrocarbons, C11-12isoalkanes,<2% aromatics

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.