Learn More
Thermo Scientific™ ThermoPrime™ TaqDNA Polymerase PCR Kts and Master Mxes
Description
Thermo Scientific™ ThermoPrime mixes are optimized for PCR applications requiring increased yield and sensitivity, to produce large amounts of PCR product.
Highlights:
- Enhanced enzyme stability for increased yield
- Advanced buffering system minimizes PCR optimization steps
- Convenient Master Mix formats save time and reduce the risk of sample contamination during assay setup
Recommended for:
Amplifying fragments up to 5kb; Routine PCR
Specifications
Specifications
| Content And Storage | Enzyme Kit. Store at -20°C for long-term storage. Stable at 4°C for up to four weeks. Avoid freeze-thaw cycles. |
| Description | ThermoPrime DNA Polymerase Enzyme Kit |
| Quantity | 400 x 25µL Rxns |
| For Use With (Application) | Standard PCR, RT-PCR, DNA Sequencing, Genotyping, Pathogen Detection |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.