missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Acetate Hydrochloride, TRC

Product Code. 30452901
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452901 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452901 Supplier Toronto Research Chemicals Supplier No. TRCT34407550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiamine Acetate Hydrochloride
CAS 1037-29-2
Purity Grade Notes HPLC
Molecular Formula C14H19N4O2S.HCl.Cl
Quantity 50 mg
Synonym Thiazolium','5-[2-(acetyloxy)ethyl]-3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-','chloride','hydrochloride (1:1:1), Thiamine','acetate (ester)','monohydrochloride (8CI), Thiamine','acetate','hydrochloride (7CI), Thiazolium','5-[2-(acetyloxy)ethyl]-3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-','chloride','monohydrochloride (9CI), Thiamine Hydrochloride Imp. G (EP) as Chloride Hydrochloride
InChI Formula InChI=1S/C14H19N4O2S.2ClH/c1-9-13(4-5-20-11(3)19)21-8-18(9)7-12-6-16-10(2)17-14(12)15;;/h6,8H,4-5,7H2,1-3H3,(H2,15,16,17);2*1H/q+1;;/p-1
SMILES Cl.[Cl-].CC(=O)OCCc1sc[n+](Cc2cnc(C)nc2N)c1C
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl acetate;chloride;hydrochloride
Molecular Weight (g/mol) 379.31
Formula Weight 378.0684
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.