missing translation for 'onlineSavingsMsg'
Learn More

Thiamine-d3 Pyrophosphate Chloride (d3-Major), TRC

Product Code. 30455770
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455770 10 mg 10mg
30450384 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30455770 Supplier Toronto Research Chemicals Supplier No. TRCT34407210MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiamine-d3 Pyrophosphate Chloride
CAS 2483831-79-2
Purity Grade Notes HPLC
Molecular Formula C12H16D3ClN4O7P2S
Quantity 10 mg
Synonym 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)thiazolium-d3 Chloride, Thiamine-d3 Trihydrogen Pyrophosphate Chloride, 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-3,5-dioxa-4,6-diphosphahex-1-yl)thiazolim-d3 Chloride P,P'-Dioxide, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)thiazolium-d3 Chloride, B-Neurox-d3, Berolase-d3, Biosyth-d3, Bioxilasi-d3, Bivitasi-d3, Co-B1-d3, Cocarbina-d3, Cocarboxil-d3, Cocarboxylase-d3, Coenbione-d3, Coxylase-d3, Diphosphothiamin-d3, Diphosphothiamine-d3, Farmaneurina-d3, Metabolase-d3, Nutrase-d3, Pyrolase-d3, Pyruvodehydrase-d3, TDP-d3, TDP (thiamin ester)-d3, TPP-d3, TPP (coenzyme), Thiamin-d3 Pyrophosphate, Thiamine-d3 Diphosphate, Thiamine-d3 Diphosphate Ester Chloride, Thiamine-d3 Diphosphoric Acid Ester Chloride, Thiamine-d3 Pyrophosphate, Thiamine-d3 Pyrophosphoric Acid Ester Chloride, Thiamine-d3 Pyrophosphoric Ester, Thiamine-PP-d3, Vitamin B1-d3 Pyrophosphate Chloride
InChI Formula InChI=1S/C12H18N4O7P2S.ClH/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);1H/i2D3;
SMILES OP(OP(OCCC1=C(C)[N+](CC2=CN=C(C([2H])([2H])[2H])N=C2N)=CS1)(O)=O)(O)=O.[Cl-]
Molecular Weight (g/mol) 463.79
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.