missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride, 100%, Cell Culture Reagent, MP Biomedicals™

Product Code. 11462041 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462041 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11462041 Supplier MP Biomedicals Supplier No. 0219474980
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 67-03-8
Molecular Formula C12H18Cl2N4OS
Quantity 100 g
Synonym thiamine hydrochloride, aneurine hydrochloride, thiamine hcl, vitamin b1 hydrochloride, thiamine dichloride, thiamine chloride hydrochloride, trophite, thiamin chloride, vitamin b1, thiaminium chloride
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Molecular Weight (g/mol) 337.26
PubChem CID 6202
ChEBI CHEBI:49105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.