missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride, Powder, MP Biomedicals

Product Code. 11453421 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453421 25 g 25g
11463421 100 g 100g
11473421 250 g 250g
11483421 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11453421 Supplier MP Biomedicals Supplier No. 0210302825
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.

Thiamine hydrochloride is used as a food-additive to add a brothy/meaty flavor to gravies or soups. It is also used as food supplement. Used in fluorometric determination of mercury.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H17ClN4OS·HCl
Molecular Weight (g/mol) 337.263
MDL Number MFCD00012780
Synonym Aneurine hydrochloride, Vitamin B1 hydrochloride, Vitamin B1 HCl, vitamin b1 mononitrate, aneurine mononitrate, aneurine nitrate, betabion mononitrate, thiamine nitrate salt, vitamin b sub 1 nitrate, thiamine mononitrate usp

Specifications

Quantity 25 g
Beilstein 3851771
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-
Formula Weight 337.3
Physical Form Powder
Chemical Name or Material Thiamine Hydrochloride, Food Supplement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.