missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride, TRC

Product Code. 17913165
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913165 10 g 10g
17950755 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17913165 Supplier Toronto Research Chemicals Supplier No. TRCT34418510G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H17N4OS.Cl.HCl
Molecular Weight (g/mol) 337.27
Synonym Thiamine Hydrochloride, Vitamin B1, Thiamine monohydrochloride, Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, chloride, hydrochloride (1:1:1), Thiamine, monohydrochloride (8CI), Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, chloride, monohydrochloride (9CI), 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methylthiazolium chloride monohydrochloride, 3-(4-Amino-2-methylpyrimidyl-5-methyl)-4-methyl-5,β-hydroxyethylthiazolium chloride hydrochloride, Aneurine hydrochloride, Apate drops, Beatine, Bedome, Begiolan, Benerva, Bequin, Berin, Betabion, Betalin S, Betaxin, Bethiazine, Beuion, Bevitine, Bewon, Biamine, Biuno, Bivatin, Bivita, Clotiamina, Eskapen, Eskaphen, Lixa-Beta, Metabolin, Slowten, THD, Thiadoxine, Thiamin chloride, Thiamin dichloride, Thiamin hydrochloride, Thiaminal, Thiamine chloride, Thiamine chloride hydrochloride, Thiamine dichloride, Thiamine hydrochloride, Thiaminium chloride, Thiaminium chloride hydrochloride, Thiamol, Thiavit, Tiamidon, Tiaminal, Trophite, Vetalin S, Vinothiam, Vitamin B1 hydrochloride, Vitaneuron
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
SMILES Cl.[Cl-].Cc1ncc(C[n+]2csc(CCO)c2C)c(N)n1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1
Formula Weight 336.0578
Percent Purity >95%
Chemical Name or Material Thiamine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.