missing translation for 'onlineSavingsMsg'
Learn More

Thian-4-one, 97%, Maybridge

Product Code. 10146922
Change view
Click to view available options
:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10146922 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10146922 Supplier Thermo Scientific Maybridge Supplier No. RF03869DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1072-72-6
Assay Percent Range 97%
Molecular Formula C5H8OS
MDL Number MFCD00006660
Quantity 1 g
Synonym tetrahydrothiopyran-4-one, tetrahydro-4h-thiopyran-4-one, 4-oxothiane, 4-thiacyclohexanone, penthianone, 4h-thiopyran-4-one, tetrahydro, 1-thiacyclohexan-4-one, dihydro-2h-thiopyran-4 3h-one, 4-oxotetrahydrothiopyran, tetrahydro-4-thiopyrone
InChI Key OVRJVKCZJCNSOW-UHFFFAOYSA-N
SMILES O=C1CCSCC1
IUPAC Name thian-4-one
Molecular Weight (g/mol) 116.18
PubChem CID 66173
Formula Weight 116.18
Percent Purity 97%
Chemical Name or Material Thian-4-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.