missing translation for 'onlineSavingsMsg'
Learn More

Thiane, TRC

Product Code. 30453144
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453144 10 g 10g
30455253 5 g 5g
30450170 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453144 Supplier Toronto Research Chemicals Supplier No. TRCT34414510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1613-51-0
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.2
Synonym NSC 9459, Pentamethylene Sulfide, Penthiophane, Tetrahydro-2H-thiopyran, Tetrahydrothiapyran, Tetrahydrothiopyran, Thiacyclohexane
IUPAC Name thiane
SMILES C1CCSCC1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H10S/c1-2-4-6-5-3-1/h1-5H2
Formula Weight 102.0503
Chemical Name or Material Thiane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.